| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008004.hg.1 |
ENST00000416951.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008004.hg.1 |
ENST00000416951.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008004.hg.1 |
ENST00000416951.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008004.hg.1 |
ENST00000416951.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008004.hg.1 |
ENST00000416951.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008004.hg.1 |
ENST00000416951.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008004.hg.1 |
ENST00000448211.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008004.hg.1 |
ENST00000448211.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008004.hg.1 |
NR_120347.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008004.hg.1 |
NR_120347.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008004.hg.1 |
NR_120347.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008004.hg.1 |
ENST00000416951.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008004.hg.1 |
ENST00000416951.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008004.hg.1 |
ENST00000416951.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008004.hg.1 |
ENST00000416951.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008004.hg.1 |
ENST00000416951.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008004.hg.1 |
ENST00000416951.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008004.hg.1 |
ENST00000448211.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008004.hg.1 |
ENST00000448211.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008004.hg.1 |
NR_120347.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008004.hg.1 |
NR_120347.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008004.hg.1 |
NR_120347.1 |
Refseq |