Assay Details
| Gene Symbol: RMST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF429306.1 | - | 4 - 5 | 664 | 116 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Downstream | microRNA 135a-2 | MIR135A2 |
MIRN135-2, MIRN135A2, mir-135a-2 |
2.8kb | Sense | Chr. 12 - 97563812 - 97563911 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000547946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000547946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000547946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000547996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000547996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000547996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000547996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000547996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000552411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000552411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000552411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000610287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000611651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000611980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000613072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000616309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000616886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000617263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000618704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000619945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000621935.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000547946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000547946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000547946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000547996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000547996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000547996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000547996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000547996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000552411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000552411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000552411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000610287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000611651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000611980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000613072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000616309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000616886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000617263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000618704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000619945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000621935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | NR_024037.1 | Refseq |

RMST
Genomic Map