Assay Details
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tumor necrosis factor superfamily member 14 | TNFSF14 |
CD258, HVEML, LIGHT, LTg |
-47.7kb | Anti-sense | Chr. 19 - 6658126 - 6670595 on NCBI Build GRCh38 |
Downstream | G protein-coupled receptor 108 | GPR108 | LUSTR2 | 11.5kb | Anti-sense | Chr. 19 - 6729914 - 6737622 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
