Assay Details
| Gene Symbol: MIRLET7I | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029661.1 | 1 | 45 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1465 | LINC01465 | C12orf61 | -252.0 | Anti-sense | Chr. 12 - 62601751 - 62603434 on NCBI Build GRCh38 |
| Downstream |
protein phosphatase, Mg2+/Mn2+ dependent 1H |
PPM1H |
ARHCL1, NERPP-2C, URCC2 |
40.2kb | Anti-sense | Chr. 12 - 62643982 - 62935135 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1200007962.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12000558.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834810 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834792 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834829 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834816 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834806 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08399714 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834793 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08399712 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834813 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834820 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834795 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834784 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834815 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08399700 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834831 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834802 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834812 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08399678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834800 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834824 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834791 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834807 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834799 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834819 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834788 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08399699 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08399689 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834817 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834828 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16565921 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834787 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834827 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834830 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834834 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834835 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16565922 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834784 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08399714 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834807 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08399689 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834810 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08399712 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834819 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834834 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834791 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834793 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08399699 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834816 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16565921 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834788 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834792 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834828 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834827 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834824 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834806 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834799 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834800 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08399700 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834815 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16565922 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834813 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834820 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834830 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834835 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834831 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834829 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834812 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08399678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834817 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834802 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834787 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28834795 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.1 ST Array | 7956737 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | ENST00000362309.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | ENST00000619323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007962.hg.1 | NR_029661.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007514.hg.1 | ENST00000362309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007514.hg.1 | NR_029661.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007514.hg.1 | TC12000558.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | ENST00000362309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | ENST00000550290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | ENST00000619323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007962.hg.1 | NR_029661.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200158371.hg.1 | ENST00000362309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200158371.hg.1 | NR_029661.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200158371.hg.1 | ENST00000362309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200158371.hg.1 | NR_029661.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200158371.hg.1 | TC1200007962.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3309204 | NR_029661.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3309204 | NR_029661.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309204 | NR_029661.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309204 | NR_029661.1 | human_transcriptome |

MIRLET7I
Genomic Map