Assay Details
| Gene Symbol: E2F6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278275.1 | NP_001265204.1 | 3 - 4 | 536 | 110 | |
| XM_017003548.1 | XP_016859037.1 | 3 - 4 | 536 | 110 | ||
| GenBank mRNA | AY083996.1 | - | 3 - 4 | 515 | 111 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 570 | LINC00570 | ncRNA-a5 | -50.5kb | Anti-sense | Chr. 2 - 11393981 - 11403077 on NCBI Build GRCh38 |
| Downstream | growth regulation by estrogen in breast cancer 1 | GREB1 | 76.9kb | Anti-sense | Chr. 2 - 11534116 - 11642788 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY083996.1, NM_001278275.1, XM_017003548.1 transcripts.
Set Membership: |
|

E2F6
Genomic Map