Assay Details
| Gene Symbol: RPSAP58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003662.3 | 4 | 390 | 135 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 681 | ZNF681 | -68.2kb | Anti-sense | Chr. 19 - 23739195 - 23758891 on NCBI Build GRCh38 | |
| Downstream | zinc finger protein 726 | ZNF726 | 86.7kb | Sense | Chr. 19 - 23914882 - 23933967 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000404.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400009753.hg.1 | ENST00000464073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008073.hg.1 | ENST00000488229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015092.hg.1 | ENST00000431241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015092.hg.1 | ENST00000431241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015092.hg.1 | ENST00000431241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010097.hg.1 | ENST00000432681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018517.hg.1 | ENST00000453374.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000311832.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000311832.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000311832.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000311832.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000311832.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000311832.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000311832.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000311832.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000311832.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000427417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000427417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000427417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000427417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000427417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000442699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000442699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000442699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000444166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000444166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000444166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000444166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000444166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000445644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000445644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000445644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000445644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000445644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000455659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000455659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000455659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000455659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000458337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000458337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000458337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000458337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000458337.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000471704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000471704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000471704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000471704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000471704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000471704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000472729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000472729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000472729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000472729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000472729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000472729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | ENST00000472729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | NM_152516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | NM_152516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | NM_152516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | NM_152516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | NM_152516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | NM_152516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | NM_152516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007739.hg.1 | NM_152516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013732.hg.1 | ENST00000445739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013732.hg.1 | ENST00000445739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013732.hg.1 | ENST00000456555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013732.hg.1 | ENST00000456555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013732.hg.1 | ENST00000456555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | ENST00000462186.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | ENST00000462186.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | ENST00000462186.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | ENST00000462186.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | ENST00000471224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | ENST00000471224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | ENST00000471224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | ENST00000471224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | ENST00000471224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | ENST00000472297.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | ENST00000472297.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | ENST00000472297.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | ENST00000472297.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | ENST00000475499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | ENST00000475499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | ENST00000484897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | ENST00000484897.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | NR_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | NR_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | NR_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | NR_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | NR_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | NR_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | NR_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011698.hg.1 | NR_003662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000311832.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000311832.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000311832.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000311832.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000311832.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000311832.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000311832.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000311832.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000311832.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000427417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000427417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000427417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000427417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000427417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000442699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000442699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000442699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000444166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000444166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000444166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000444166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000444166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000445644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000445644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000445644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000445644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000445644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000455659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000455659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000455659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000455659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000458337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000458337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000458337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000458337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000458337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000471704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000471704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000471704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000471704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000471704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000471704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000472729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000472729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000472729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000472729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000472729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000472729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | ENST00000472729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | NM_152516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | NM_152516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | NM_152516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | NM_152516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | NM_152516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | NM_152516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | NM_152516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007739.hg.1 | NM_152516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015092.hg.1 | ENST00000431241.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015092.hg.1 | ENST00000431241.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015092.hg.1 | ENST00000431241.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018517.hg.1 | ENST00000453374.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013732.hg.1 | ENST00000445739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013732.hg.1 | ENST00000445739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013732.hg.1 | ENST00000456555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013732.hg.1 | ENST00000456555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013732.hg.1 | ENST00000456555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010097.hg.1 | ENST00000432681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008073.hg.1 | ENST00000488229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009753.hg.1 | ENST00000464073.1 | Ensembl |

RPSAP58
Genomic Map