Assay Details
Gene Symbol: SNORD116-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003316.1 | 1 | 42 | 90 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC15000048.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4172 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16760219 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30023289 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543423 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543434 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543425 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30023273 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30023288 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30023279 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543421 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543428 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543432 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30023282 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30023273 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543421 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543425 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30023282 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543432 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543434 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16760219 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543423 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30023288 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30023279 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543428 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30023289 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543517 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543485 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543505 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543490 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543462 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543508 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543481 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16760238 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543430 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543456 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543506 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543426 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543436 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543432 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543492 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543424 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543420 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543495 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543469 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543484 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543474 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543515 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543473 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543467 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543489 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543504 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543503 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543512 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543435 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543460 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543422 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543479 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543428 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543431 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543457 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NR_003316_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7981954 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7981959 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7981967 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7981963 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7981950 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7981958 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7981962 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7981966 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7981953 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7981949 | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1500006573.hg.1 | ENST00000384335.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006573.hg.1 | NR_003316.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15000606.hg.1 | ENST00000384335.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15000606.hg.1 | NR_003316.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15000606.hg.1 | TC15000048.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1500006573.hg.1 | ENST00000384335.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006573.hg.1 | NR_003316.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145725.hg.1 | ENST00000384335.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145725.hg.1 | NR_003316.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145725.hg.1 | ENST00000384335.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145725.hg.1 | NR_003316.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145725.hg.1 | TC1500006573.hg.1 | Clariom D TC |
