Assay Details
Gene Symbol: LINC00696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027331.1 | 1 | 1849 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | dual specificity phosphatase 7 | DUSP7 |
MKPX, PYST2 |
-5.6kb | Sense | Chr. 3 - 52048919 - 52056445 on NCBI Build GRCh38 |
Downstream | POC1 centriolar protein A | POC1A |
PIX2, SOFT, WDR51A |
10.1kb | Sense | Chr. 3 - 52075233 - 52154690 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03001446.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 139862 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3335614 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5117 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5116 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 21181 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3335614 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3335614 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22372652 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237791 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237789 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15438448 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237783 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15438431 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15438437 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22372693 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237788 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237749 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237764 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237776 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237800 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22372637 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22372639 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237759 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237807 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237747 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237774 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22372672 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237761 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237779 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237758 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237805 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237754 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237771 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15438428 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237773 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237743 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22372677 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237777 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237781 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237796 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237802 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237744 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237790 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237750 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237755 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15438434 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237751 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237757 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15438430 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22372650 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237746 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237786 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237804 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237785 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15438444 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22372622 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237809 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237780 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22372668 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22372693 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22372652 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237758 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237761 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237757 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15438448 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237743 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237749 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237764 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237790 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237796 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237791 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237802 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237788 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237754 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15438434 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237781 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237759 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237746 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237804 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22372672 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22372637 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237771 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15438444 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15438430 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237786 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237773 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237747 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237777 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237789 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237776 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237780 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22372639 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237779 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237755 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15438437 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237800 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22372668 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237750 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237805 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237807 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22372677 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237744 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237783 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15438428 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237774 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237785 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15438431 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237809 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02237751 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22372650 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22372622 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00704174 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237723 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237708 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237750 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237749 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15438437 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15438448 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237791 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237790 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237776 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237780 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237744 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237683 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237786 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237757 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237802 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237701 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237793 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237765 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237696 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237725 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237706 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15438450 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237800 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237689 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237727 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237774 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15438403 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237712 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237747 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237777 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237684 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15438425 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237807 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237805 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237715 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237763 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237758 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237773 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237761 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237783 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237785 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237812 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15438434 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15438430 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237771 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237672 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237746 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15438405 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237703 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15438444 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237687 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237751 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15438453 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237754 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15438400 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237676 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237752 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237674 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237781 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237801 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237815 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237710 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237700 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237692 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237755 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237695 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237720 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237817 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15438407 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237704 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237680 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237768 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237729 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237717 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237788 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237809 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237678 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02237698 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15438436 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03026154.hg.1 | TC03001446.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03026154.hg.1 | NR_027331.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03026155.hg.1 | TC03001446.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03026155.hg.1 | ENST00000541313.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026155.hg.1 | NR_027331.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03026156.hg.1 | TC03001446.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03026156.hg.1 | NR_027331.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183211.hg.1 | NR_027331.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183211.hg.1 | NR_027331.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183211.hg.1 | TC0300011153.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021130 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021130 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3240168 | NR_027331.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3240168 | NR_027331.1 | human_transcriptome |
