Assay Details
| Gene Symbol: FOXI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135649.1 | NP_001129121.1 | 2 | 953 | 126 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002073.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 128485 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 247305 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 614009 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P15391 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 25896 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 15027 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P15739367 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P57923494 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P15391 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P15391 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426190 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337700 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15271492 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426195 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337661 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337680 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426206 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337668 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15271491 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426207 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15271504 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426217 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337656 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426182 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426204 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337671 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337657 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426180 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426186 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426191 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337696 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337662 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426202 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426214 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337692 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426198 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15271486 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15271488 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337679 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337691 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337659 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426200 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337701 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337658 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426208 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426213 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426180 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337661 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15271491 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15271486 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426213 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337696 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15271504 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337680 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337657 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426186 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426182 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426217 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15271488 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337662 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337691 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337701 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15271492 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337659 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337668 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426207 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337700 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426204 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426190 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337658 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426208 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426200 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337671 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337656 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426191 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426206 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426195 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426214 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337679 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426198 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337692 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01337673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21426202 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P15391 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P15391 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337605 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337687 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337677 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337666 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15271481 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337601 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337651 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337650 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337639 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15271485 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337600 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337612 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337656 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337699 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337646 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337626 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337628 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337689 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15271488 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337672 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337692 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15271503 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15271501 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337634 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337670 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337602 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337624 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337604 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337637 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337649 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337641 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337630 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337622 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337696 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337661 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337674 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337683 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337659 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337694 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15271504 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337643 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337681 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15271499 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15271491 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337701 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337679 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337654 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337620 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337668 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337608 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337662 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337619 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337610 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15271497 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337704 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337652 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337615 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337645 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01337664 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P15739367 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Genome U219 Array | 11741892_at | - | - |
| Affymetrix Human Genome U219 Array | 11741892_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033196.hg.1 | TC02002073.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033196.hg.1 | NM_001135649.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013373.hg.1 | ENST00000398142.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013373.hg.1 | ENST00000398142.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013373.hg.1 | ENST00000398142.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013373.hg.1 | ENST00000428390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013373.hg.1 | ENST00000428390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013373.hg.1 | ENST00000428390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013373.hg.1 | ENST00000428390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013373.hg.1 | ENST00000428390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013373.hg.1 | ENST00000428390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013373.hg.1 | ENST00000428390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013373.hg.1 | NM_001135649.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013373.hg.1 | NM_001135649.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033195.hg.1 | TC02002073.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033195.hg.1 | NM_001135649.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033198.hg.1 | TC02002073.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033198.hg.1 | NM_001135649.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200197638.hg.1 | ENST00000398142.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200197638.hg.1 | ENST00000428390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200197638.hg.1 | ENST00000428390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200197638.hg.1 | NM_001135649.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200197638.hg.1 | ENST00000398142.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200197638.hg.1 | ENST00000428390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200197638.hg.1 | ENST00000428390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200197638.hg.1 | NM_001135649.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200197638.hg.1 | TC0200013373.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200013373.hg.1 | ENST00000398142.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013373.hg.1 | ENST00000398142.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013373.hg.1 | ENST00000398142.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013373.hg.1 | ENST00000428390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013373.hg.1 | ENST00000428390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013373.hg.1 | ENST00000428390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013373.hg.1 | ENST00000428390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013373.hg.1 | ENST00000428390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013373.hg.1 | ENST00000428390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013373.hg.1 | ENST00000428390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013373.hg.1 | NM_001135649.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013373.hg.1 | NM_001135649.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200197639.hg.1 | ENST00000398142.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200197639.hg.1 | ENST00000428390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200197639.hg.1 | ENST00000428390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200197639.hg.1 | NM_001135649.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200197639.hg.1 | ENST00000398142.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200197639.hg.1 | ENST00000428390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200197639.hg.1 | ENST00000428390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200197639.hg.1 | NM_001135649.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200197639.hg.1 | TC0200013373.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A013984 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A013984 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 164328 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3245306 | NM_001135649.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1726940 | - | - |
| HumanHT-12 V4.0 expression beadchip | ILMN_3245306 | NM_001135649.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3245306 | NM_001135649.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3245306 | NM_001135649.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1726940 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1726940 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1726940 | - | - |

FOXI3
Genomic Map