Assay Details
| Gene Symbol: RPL23AP32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002229.2 | 1 | 49 | 78 | ||
| GenBank mRNA | BC096706.1 | - | 1 | 49 | 78 | |
| BC096740.1 | - | 1 | 49 | 78 | ||
| BC098172.1 | - | 1 | 49 | 78 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 2 open reading frame 73 | C2orf73 | -145.5kb | Sense | Chr. 2 - 54329961 - 54383673 on NCBI Build GRCh38 | |
| Downstream | echinoderm microtubule associated protein like 6 | EML6 | 193.8kb | Sense | Chr. 2 - 54723467 - 54972908 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000317.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P108932 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P379649 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P108932 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 6960 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 27403 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 5802 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 27494 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 25191 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 42654 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 10990 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 40475 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 32020 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 55859 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 48078 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 15868 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P108932 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P108932 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189852 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15244612 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270462 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189848 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189872 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189881 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270489 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189892 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189876 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270483 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270494 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270454 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270513 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189865 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189841 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270490 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270499 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270460 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270451 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189883 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270510 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15244614 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189835 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270459 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270503 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189869 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15244608 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270504 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270480 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270506 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189836 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189863 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189871 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270478 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189891 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189851 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189873 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270475 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189867 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189844 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270470 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270498 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270476 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15244608 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189863 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270462 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270480 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270476 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189872 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189869 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189881 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270460 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189848 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270494 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270506 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189891 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189851 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189871 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189883 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270454 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270498 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270475 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270490 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189873 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270451 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270470 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270513 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270503 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189841 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189844 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189835 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270483 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189876 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15244612 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270478 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189852 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189836 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15244614 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189867 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270489 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189892 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270510 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270499 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270459 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01189865 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21270504 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P108932 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P379649 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P379649 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P108932 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15244609 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189838 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189846 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189876 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15244615 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189888 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189849 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189880 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189870 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189862 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189874 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189858 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189865 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189844 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189836 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15244606 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189886 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189851 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189884 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189867 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189890 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189835 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189841 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189840 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15244608 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189864 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189868 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189872 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15244612 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01189856 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_002229_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8042039 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8042038 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 207283_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004851.hg.1 | TC02000317.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004851.hg.1 | NR_002229.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016801.hg.1 | ENST00000437556.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007611.hg.1 | ENST00000395315.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007611.hg.1 | NR_002229.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007611.hg.1 | NR_002229.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007611.hg.1 | NR_002229.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200152602.hg.1 | ENST00000395315.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200152602.hg.1 | NR_002229.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200152602.hg.1 | ENST00000395315.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200152602.hg.1 | NR_002229.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200152602.hg.1 | TC0200007611.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100016801.hg.1 | ENST00000437556.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200152601.hg.1 | NR_002229.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200152601.hg.1 | NR_002229.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200152601.hg.1 | TC0200007611.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200007611.hg.1 | ENST00000395315.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007611.hg.1 | NR_002229.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007611.hg.1 | NR_002229.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007611.hg.1 | NR_002229.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200152603.hg.1 | NR_002229.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200152603.hg.1 | NR_002229.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200152603.hg.1 | TC0200007611.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 207283_at | - | - |
| Human Genome U133 Plus 2.0 Array | 207283_PM_at | - | - |
| Human Genome U133A 2.0 Array | 207283_PM_at | - | - |
| Human Genome U133A 2.0 Array | 207283_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NR_002229_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw552514 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207283_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207283_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207283_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207283_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207283_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207283_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207283_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207283_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207283_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207283_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207283_at1 | - | - |
| Human Genome U133 Set | 207283_PM_at | - | - |
| Human Genome U133 Set | 207283_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015661 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015661 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_2124757 | BC096706.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2124757 | BC096740.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2124757 | BC098172.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2124757 | NR_002229.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1752897 | - | - |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1752897 | - | - |
| HumanHT-12 V4.0 expression beadchip | ILMN_2124757 | BC096706.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2124757 | BC096740.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2124757 | BC098172.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2124757 | NR_002229.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1752897 | - | - |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2124757 | BC096706.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2124757 | BC096740.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2124757 | BC098172.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2124757 | NR_002229.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2124757 | BC096706.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2124757 | BC096740.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2124757 | BC098172.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2124757 | NR_002229.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1752897 | - | - |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 7650273 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_20679 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2124757 | BC096706.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2124757 | BC096740.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2124757 | BC098172.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2124757 | NR_002229.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1752897 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2124757 | BC096706.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2124757 | BC096740.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2124757 | BC098172.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2124757 | NR_002229.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2124757 | BC096706.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2124757 | BC096740.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2124757 | BC098172.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2124757 | NR_002229.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1752897 | - | - |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1752897 | - | - |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2124757 | BC096706.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2124757 | BC096740.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2124757 | BC098172.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2124757 | NR_002229.2 | human_transcriptome |

RPL23AP32
Genomic Map