Assay Details
Gene Symbol: LINC00470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_023927.1 | 1 - 2 | 244 | 109 | ||
GenBank mRNA | AF295730.1 | - | 1 - 2 | 266 | 109 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC388456 | LOC388456 | -430.7kb | Sense | Chr. 18 - 926083 - 928129 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC101926997 | LOC101926997 | 1.2mb | Sense | Chr. 18 - 2517250 - 2519260 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
