Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT HG-U133+ PM Array Plate |
1556661_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009016.hg.1 |
ENST00000617229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009016.hg.1 |
ENST00000623117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009016.hg.1 |
ENST00000623117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009016.hg.1 |
ENST00000623117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009016.hg.1 |
NR_024009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009016.hg.1 |
NR_024009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009016.hg.1 |
NR_024009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014916.hg.1 |
NR_024009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014916.hg.1 |
TC22000888.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200165324.hg.1 |
TC2200009016.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200165324.hg.1 |
ENST00000623117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200165324.hg.1 |
NR_024009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200165324.hg.1 |
ENST00000623117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200165324.hg.1 |
NR_024009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200165326.hg.1 |
TC2200009016.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200165326.hg.1 |
ENST00000623117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200165326.hg.1 |
NR_024009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200165326.hg.1 |
ENST00000623117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200165326.hg.1 |
NR_024009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200165325.hg.1 |
TC2200009016.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200165325.hg.1 |
ENST00000617229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200165325.hg.1 |
ENST00000623117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200165325.hg.1 |
NR_024009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200165325.hg.1 |
ENST00000617229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200165325.hg.1 |
ENST00000623117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200165325.hg.1 |
NR_024009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009016.hg.1 |
ENST00000617229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009016.hg.1 |
ENST00000623117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009016.hg.1 |
ENST00000623117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009016.hg.1 |
ENST00000623117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009016.hg.1 |
NR_024009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009016.hg.1 |
NR_024009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009016.hg.1 |
NR_024009.1 |
Refseq |
Human Genome U133 Plus 2.0 Array |
1556661_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1556661_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1556661_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1556661_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1556661_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1556661_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1556661_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1556661_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1556661_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1556661_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1556661_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1556661_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1556661_at2 |
- |
- |