Assay Details
| Gene Symbol: ADGRE4P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024075.1 | 9 - 10 | 1049 | 83 | ||
| GenBank mRNA | AF489700.1 | - | 9 - 10 | 1049 | 83 | |
| AY181245.1 | - | 5 - 6 | 558 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | adhesion G protein-coupled receptor E1 | ADGRE1 |
EMR1, TM7LN3 |
-30.7kb | Anti-sense | Chr. 19 - 6887549 - 6940453 on NCBI Build GRCh38 |
| Downstream | microtubule affinity regulating kinase 1 pseudogene | FLJ25758 | 32.5kb | Sense | Chr. 19 - 7004953 - 7006415 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001106.hg.1
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19015236.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015236.hg.1 | TC19001106.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900176086.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176086.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176086.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900176086.hg.1 | TC1900009452.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900176086.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176086.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176086.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000597372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | ENST00000600751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009452.hg.1 | NR_024075.1 | Refseq |

ADGRE4P
Genomic Map