Assay Details
Gene Symbol: ZFP91-CNTF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_024091.1 | 11 - 12 | 1740 | 96 | ||
GenBank mRNA | AB057443.1 | - | 11 - 12 | 1686 | 96 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ZFP91 zinc finger protein | ZFP91 |
DMS-8, DSM-8, FKSG11, PZF, ZFP-91, ZNF757 |
Sense | Chr. 11 - 58579114 - 58621550 on NCBI Build GRCh38 | |
Downstream | glycine-N-acyltransferase | GLYAT |
ACGNAT, CAT, GAT |
85.9kb | Anti-sense | Chr. 11 - 58708757 - 58731974 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11000477.hg.1
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11005809.hg.1 | ENST00000361987.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11005809.hg.1 | ENST00000389919.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11005809.hg.1 | NM_000614.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11005809.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11005809.hg.1 | TC11000477.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11005797.hg.1 | ENST00000316059.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11005797.hg.1 | ENST00000389918.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11005797.hg.1 | ENST00000389919.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11005797.hg.1 | NM_001197051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11005797.hg.1 | NM_053023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11005797.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11005797.hg.1 | TC11000477.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11005795.hg.1 | ENST00000316059.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11005795.hg.1 | ENST00000389918.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11005795.hg.1 | ENST00000389919.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11005795.hg.1 | NM_001197051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11005795.hg.1 | NM_053023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11005795.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11005795.hg.1 | TC11000477.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11005796.hg.1 | ENST00000316059.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11005796.hg.1 | ENST00000389918.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11005796.hg.1 | ENST00000389919.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11005796.hg.1 | NM_001197051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11005796.hg.1 | NM_053023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11005796.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11005796.hg.1 | TC11000477.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100200564.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200564.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200564.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100200564.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200564.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200564.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100200564.hg.1 | TC1100013007.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100200565.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200565.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200565.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100200565.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200565.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200565.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100200565.hg.1 | TC1100013007.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100200568.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200568.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200568.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100200568.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200568.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200568.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100200568.hg.1 | TC1100013007.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100200566.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200566.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200566.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100200566.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200566.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200566.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100200566.hg.1 | TC1100013007.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000389919.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | ENST00000422974.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | NR_024091.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013007.hg.1 | NR_024091.1 | Refseq |
