Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011915.hg.1 |
TC09000896.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011915.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011915.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011915.hg.1 |
NM_001243203.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011915.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
ENST00000485372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
ENST00000485372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
ENST00000485372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
ENST00000623170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
ENST00000623170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
ENST00000623170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
NM_001243203.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
NM_001243203.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
NM_001243203.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
NM_001243203.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
NM_001243203.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009479.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011919.hg.1 |
TC09000896.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011919.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011919.hg.1 |
ENST00000485372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011919.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011919.hg.1 |
NM_001243203.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011919.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011916.hg.1 |
TC09000896.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011916.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011916.hg.1 |
ENST00000485372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011916.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011916.hg.1 |
NM_001243203.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011916.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011918.hg.1 |
TC09000896.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011918.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011918.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09011918.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165038.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165038.hg.1 |
ENST00000485372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165038.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165038.hg.1 |
NM_001243203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165038.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165038.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165038.hg.1 |
ENST00000485372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165038.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165038.hg.1 |
NM_001243203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165038.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165038.hg.1 |
TC0900009479.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900165039.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165039.hg.1 |
ENST00000485372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165039.hg.1 |
ENST00000623170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165039.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165039.hg.1 |
NM_001243203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165039.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165039.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165039.hg.1 |
ENST00000485372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165039.hg.1 |
ENST00000623170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165039.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165039.hg.1 |
NM_001243203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165039.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165039.hg.1 |
TC0900009479.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900165041.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165041.hg.1 |
ENST00000623170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165041.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165041.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165041.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165041.hg.1 |
ENST00000623170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165041.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165041.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165041.hg.1 |
TC0900009479.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
ENST00000485372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
ENST00000485372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
ENST00000485372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
ENST00000623170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
ENST00000623170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
ENST00000623170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
NM_001243203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
NM_001243203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
NM_001243203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
NM_001243203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
NM_001243203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009479.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165037.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165037.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165037.hg.1 |
NM_001243203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165037.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165037.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165037.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165037.hg.1 |
NM_001243203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165037.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165037.hg.1 |
TC0900009479.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900165042.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165042.hg.1 |
ENST00000485372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165042.hg.1 |
ENST00000623170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165042.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165042.hg.1 |
NM_001243203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165042.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165042.hg.1 |
ENST00000259569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165042.hg.1 |
ENST00000485372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165042.hg.1 |
ENST00000623170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165042.hg.1 |
NM_001243202.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165042.hg.1 |
NM_001243203.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165042.hg.1 |
NM_012416.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165042.hg.1 |
TC0900009479.hg.1 |
Clariom D TC |