Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047172.hg.1 |
TC01003024.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047172.hg.1 |
ENST00000369492.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047172.hg.1 |
NR_024125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
ENST00000369491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
ENST00000369491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
ENST00000369491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
ENST00000369492.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
ENST00000369492.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
ENST00000369492.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
ENST00000493908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
ENST00000493908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
ENST00000493908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
ENST00000608511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
ENST00000608511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
ENST00000608511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
ENST00000608511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
ENST00000608511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
ENST00000608511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
ENST00000608511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
ENST00000608511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
ENST00000608511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
ENST00000608511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
NR_024124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
NR_024124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
NR_024124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
NR_024124.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
NR_024125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
NR_024125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
NR_024125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
NR_024125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
NR_024125.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
NR_024126.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
NR_024126.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
NR_024126.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
NR_024126.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
NR_027645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
NR_027645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
NR_027645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
NR_027646.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
NR_027646.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
NR_027646.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015391.hg.1 |
NR_027646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215007.hg.1 |
ENST00000369492.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215007.hg.1 |
NR_024125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215007.hg.1 |
ENST00000369492.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215007.hg.1 |
NR_024125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215007.hg.1 |
TC0100015391.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
ENST00000369491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
ENST00000369491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
ENST00000369491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
ENST00000369492.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
ENST00000369492.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
ENST00000369492.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
ENST00000493908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
ENST00000493908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
ENST00000493908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
ENST00000608511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
ENST00000608511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
ENST00000608511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
ENST00000608511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
ENST00000608511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
ENST00000608511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
ENST00000608511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
ENST00000608511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
ENST00000608511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
ENST00000608511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
NR_024124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
NR_024124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
NR_024124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
NR_024124.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
NR_024125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
NR_024125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
NR_024125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
NR_024125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
NR_024125.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
NR_024126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
NR_024126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
NR_024126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
NR_024126.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
NR_027645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
NR_027645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
NR_027645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
NR_027646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
NR_027646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
NR_027646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015391.hg.1 |
NR_027646.1 |
Refseq |