Assay Details
Gene Symbol: GHRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302825.1 | NP_001289754.1 | 1 - 2 | 102 | 100 | |
GenBank mRNA | EF549566.1 | - | 1 - 2 | 93 | 100 | |
EF549573.1 | - | 1 - 2 | 72 | 100 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ghrelin opposite strand/antisense RNA | GHRLOS |
GHRL-AS1, GHRLAS, NCRNA00068 |
Anti-sense | Chr. 3 - 10280952 - 10293449 on NCBI Build GRCh38 | |
Downstream |
SEC13 homolog, nuclear pore and COPII coat complex component |
SEC13 |
D3S1231E, SEC13L1, SEC13R, npp-20 |
8.0kb | Sense | Chr. 3 - 10300929 - 10321188 on NCBI Build GRCh38 |

More Information
Additional Information:
This assay is in the 5'UTR region of the EF549573.1, NM_001302825.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
