| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
7906776 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
7906775 |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176306.hg.1 |
NR_024151.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176306.hg.1 |
NR_024151.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100176306.hg.1 |
TC0100010362.hg.1 |
Clariom D TC |
| Human Genome U95 Set |
56711_s_at |
- |
- |
| Human Genome U95 Set |
56711_PM_s_at |
- |
- |