Assay Details
Gene Symbol: YWHAEP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_024178.1 | 1 - 2 | 57 | 91 | ||
GenBank mRNA | AK094896.1 | - | 1 - 2 | 57 | 91 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | HNF1 homeobox B | HNF1B |
FJHN, HNF-1-beta, HNF-1B, HNF1beta, HNF2, HPC11, LF-B3, LFB3, MODY5, TCF-2, TCF2, VHNF1 |
-138.9kb | Sense | Chr. 17 - 37686431 - 37745078 on NCBI Build GRCh38 |
Downstream | TBC1 domain family member 3K | TBC1D3K | 39.1kb | Anti-sense | Chr. 17 - 37923805 - 37935371 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001412.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK094896.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17018767.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17018767.hg.1 | TC17001412.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | ENST00000617508.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | ENST00000617508.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | ENST00000617508.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | ENST00000617508.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | ENST00000617508.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | ENST00000622054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | ENST00000622054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | ENST00000622054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | ENST00000622054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010507.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000425159.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000425159.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000425159.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000425159.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000425159.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000425159.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000425159.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000572695.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000572695.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000572695.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000572695.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000573318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000573318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000573318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000573318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000573318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000575291.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000575291.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000575291.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000575291.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000575291.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000575291.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000575291.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000575291.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000575291.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | ENST00000575291.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | NR_033934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | NR_033934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | NR_033934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | NR_033934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | NR_033934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | NR_033934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | NR_033934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | NR_033934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | NR_033934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010950.hg.1 | NR_033934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17018764.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17018764.hg.1 | TC17001412.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17018766.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17018766.hg.1 | TC17001412.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000425159.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000425159.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000425159.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000425159.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000425159.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000425159.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000425159.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000572695.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000572695.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000572695.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000572695.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000573318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000573318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000573318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000573318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000573318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000575291.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000575291.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000575291.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000575291.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000575291.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000575291.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000575291.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000575291.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000575291.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | ENST00000575291.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | NR_033934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | NR_033934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | NR_033934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | NR_033934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | NR_033934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | NR_033934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | NR_033934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | NR_033934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | NR_033934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010950.hg.1 | NR_033934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | ENST00000617508.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | ENST00000617508.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | ENST00000617508.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | ENST00000617508.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | ENST00000617508.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | ENST00000622054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | ENST00000622054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | ENST00000622054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | ENST00000622054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010507.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700182673.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700182673.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700182673.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700182673.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700182673.hg.1 | TC1700010507.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700182674.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700182674.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700182674.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700182674.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700182674.hg.1 | TC1700010507.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700182671.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700182671.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700182671.hg.1 | ENST00000620903.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700182671.hg.1 | NR_024178.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700182671.hg.1 | TC1700010507.hg.1 | Clariom D TC |
