Assay Details
| Gene Symbol: SNORA30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002966.1 | 1 | 62 | 86 | ||
| GenBank mRNA | AJ609440.1 | - | 1 | 62 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC730183 | LOC730183 | -12.0kb | Anti-sense | Chr. 16 - 30697704 - 30698489 on NCBI Build GRCh38 | |
| Downstream | transmembrane protein 265 | TMEM265 | IFITMD8 | 29.5kb | Sense | Chr. 16 - 30740191 - 30745196 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000357.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3968 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3969 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514231 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984191 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16879207 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984174 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984181 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514204 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16879215 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984166 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514237 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984172 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514218 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984180 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984170 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514227 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984175 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514217 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984165 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514201 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514225 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16879210 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16879208 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16879214 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16879206 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984171 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984163 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514199 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16879219 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984167 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514214 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514216 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984169 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16879220 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984187 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16879218 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984181 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16879215 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16879214 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984191 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514204 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16879210 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984170 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16879208 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16879206 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984172 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984180 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984174 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984167 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514225 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16879218 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16879220 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514218 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514214 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984187 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514231 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514237 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514217 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514216 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984166 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514227 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16879219 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984165 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984171 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984169 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984163 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514201 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16879207 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09984175 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30514199 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09984180 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09984185 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16879212 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09984184 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16879208 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09984191 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09984178 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16879218 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09984164 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09984166 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09984165 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09984182 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16879220 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09984170 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09984161 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09984162 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09984169 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09984187 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09984175 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16879210 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09984174 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_002966_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7994927 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7994926 | - | - |
| Affymetrix Human Genome U219 Array | 11757161_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11757160_at | - | - |
| Affymetrix Human Genome U219 Array | 11757160_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11757161_PM_x_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007487.hg.1 | ENST00000384028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007487.hg.1 | NR_002966.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16005201.hg.1 | ENST00000384028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16005201.hg.1 | NR_002966.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16005201.hg.1 | TC16000357.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1600007487.hg.1 | ENST00000384028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007487.hg.1 | NR_002966.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600154706.hg.1 | ENST00000384028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600154706.hg.1 | NR_002966.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600154706.hg.1 | ENST00000384028.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600154706.hg.1 | NR_002966.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600154706.hg.1 | TC1600007487.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3238259 | NR_002966.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3238259 | AJ609440.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3238259 | NR_002966.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3238259 | AJ609440.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3238259 | NR_002966.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3238259 | AJ609440.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3238259 | NR_002966.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3238259 | AJ609440.1 | human_transcriptome |

SNORA30
Genomic Map