Assay Details
| Gene Symbol: LOC100130857 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK097050.1 | - | 1 | 2654 | 90 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | OTU deubiquitinase 7A | OTUD7A |
C15orf16, C16ORF15, CEZANNE2, OTUD7 |
-104.7kb | Anti-sense | Chr. 15 - 31483126 - 31655339 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105370940 | LOC105370940 | 50.3kb | Anti-sense | Chr. 15 - 32028534 - 32030596 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566789 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053929 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566808 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566721 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566730 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566826 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16772754 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053872 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566708 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566816 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053841 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566812 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566815 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053906 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053861 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566796 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566712 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566769 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566797 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053840 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16772750 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16772747 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566799 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566824 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566745 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053870 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566813 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053853 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566780 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053851 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16772734 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566811 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566709 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053931 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566693 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16772733 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053896 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566743 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566820 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566706 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053943 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566786 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053839 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053854 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566825 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053860 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053843 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566698 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566717 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053904 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053934 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566728 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053866 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566739 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566711 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566722 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053884 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566761 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053946 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566805 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053957 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053859 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566744 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566696 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566821 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566772 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566766 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16772753 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053948 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566753 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566718 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053835 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566762 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566734 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053878 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053842 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16772749 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566751 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566778 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16772721 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566725 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566790 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053926 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566699 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053834 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566737 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566716 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053844 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16772720 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053953 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053869 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053944 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566802 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053847 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566752 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566773 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566746 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053922 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053901 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566750 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566798 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053924 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566792 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053891 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053917 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566733 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566714 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053925 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566756 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053898 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053833 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053914 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566793 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09566779 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053939 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30053960 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 624961 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3280346 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 60099 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3280346 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P863093 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7982340 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7982339 | - | - |

LOC100130857
Genomic Map