Assay Details
| Gene Symbol: GRK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK097099.1 | - | 1 | 2553 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Downstream | regulator of G-protein signaling 10 | RGS10 | 40.1kb | Anti-sense | Chr. 10 - 119499813 - 119542752 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

GRK5
Genomic Map