Assay Details
Gene Symbol: PCOLCE-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038910.1 | 1 - 2 | 206 | 82 | ||
GenBank mRNA | AK097705.1 | - | 1 - 2 | 206 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | F-box protein 24 | FBXO24 | FBX24 | -1.2kb | Anti-sense | Chr. 7 - 100582490 - 100601117 on NCBI Build GRCh38 |
Downstream | motile sperm domain containing 3 | MOSPD3 |
CDS3, NET30 |
8.1kb | Anti-sense | Chr. 7 - 100612102 - 100615380 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001673.hg.1
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR07025796.hg.1 | TC07001673.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07025796.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07025796.hg.1 | ENST00000446022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07025796.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07025795.hg.1 | TC07001673.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07025795.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07025795.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07025791.hg.1 | TC07001673.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07025791.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07025791.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07025794.hg.1 | TC07001673.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07025794.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07025794.hg.1 | ENST00000446022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07025794.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07025792.hg.1 | TC07001673.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07025792.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07025792.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07025792.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07025793.hg.1 | TC07001673.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07025793.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07025793.hg.1 | ENST00000446022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07025793.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07025793.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000446022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000446022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000446022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000446022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700186230.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186230.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700186230.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186230.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700186230.hg.1 | TC0700012009.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000446022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000446022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000446022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000446022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012009.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700186233.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186233.hg.1 | ENST00000446022.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186233.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700186233.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186233.hg.1 | ENST00000446022.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186233.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700186233.hg.1 | TC0700012009.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700186235.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186235.hg.1 | ENST00000446022.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186235.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700186235.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186235.hg.1 | ENST00000446022.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186235.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700186235.hg.1 | TC0700012009.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700186231.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186231.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186231.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700186231.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186231.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186231.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700186231.hg.1 | TC0700012009.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700186232.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186232.hg.1 | ENST00000446022.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186232.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186232.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700186232.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186232.hg.1 | ENST00000446022.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186232.hg.1 | ENST00000544873.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186232.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700186232.hg.1 | TC0700012009.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700186234.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186234.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700186234.hg.1 | ENST00000442166.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186234.hg.1 | NR_038910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700186234.hg.1 | TC0700012009.hg.1 | Clariom D TC |
