Assay Details
| Gene Symbol: TMCO5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046005.1 | 7 - 8 | 630 | 100 | ||
| GenBank mRNA | AK098530.1 | - | 2 - 3 | 126 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | formin 1 | FMN1 |
FMN, LD |
-40.8kb | Sense | Chr. 15 - 32765544 - 33194765 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101928134 | LOC101928134 | 62.1kb | Sense | Chr. 15 - 33303657 - 33310659 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001185.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3350858 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000528283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000528283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010489.hg.1 | ENST00000529696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010489.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010489.hg.1 | TC15001185.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010492.hg.1 | ENST00000529696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010492.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010492.hg.1 | TC15001185.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010491.hg.1 | ENST00000529696.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010491.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010491.hg.1 | TC15001185.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1500164632.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164632.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164632.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164632.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164632.hg.1 | TC1500008962.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500164638.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164638.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164638.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164638.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164638.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164638.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164638.hg.1 | TC1500008962.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500164634.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164634.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164634.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164634.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164634.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164634.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164634.hg.1 | TC1500008962.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000528283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000528283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164636.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164636.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164636.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164636.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164636.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164636.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164636.hg.1 | TC1500008962.hg.1 | Clariom D TC |

TMCO5B
Genomic Map