Assay Details
| Gene Symbol: EIF4EBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC015007.1 | - | 1 | 1356 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat containing 20 | LRRC20 | -21.2kb | Sense | Chr. 10 - 70298970 - 70382650 on NCBI Build GRCh38 | |
| Downstream | nodal growth differentiation factor | NODAL | HTX5 | 26.6kb | Sense | Chr. 10 - 70431936 - 70441709 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

EIF4EBP2
Genomic Map