Assay Details
Gene Symbol: FUBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC015184.1 | - | 1 | 1253 | 111 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC100272217 | LOC100272217 | -41.2kb | Anti-sense | Chr. 9 - 130577350 - 130579494 on NCBI Build GRCh38 | |
Downstream | microRNA 6856 | MIR6856 | hsa-mir-6856 | 4.0kb | Sense | Chr. 9 - 130626297 - 130626363 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
