Assay Details
| Gene Symbol: TCEB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC019949.1 | - | 1 | 2352 | 69 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | CR991543.1 | - | 1 | Chr. 1 - 23746289 - 23747079 [+] on Build GRCh38 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal protein L11 | RPL11 |
DBA7, GIG34, L11 |
-50.3kb | Anti-sense | Chr. 1 - 23691779 - 23696425 on NCBI Build GRCh38 |
| Downstream | TCEB3 antisense RNA 1 | TCEB3-AS1 | 11.3kb | Sense | Chr. 1 - 23760382 - 23778297 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TCEB3
Genomic Map