Assay Details
| Gene Symbol: TSC22D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC036355.1 | - | 1 | 1452 | 167 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1214 | LINC01214 | -84.4kb | Sense | Chr. 3 - 150265407 - 150323747 on NCBI Build GRCh38 | |
| Downstream | stress-associated endoplasmic reticulum protein 1 | SERP1 | RAMP4 | 132.3kb | Sense | Chr. 3 - 150541993 - 150603177 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TSC22D2
Genomic Map