Assay Details
Gene Symbol: LOC284933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038917.1 | 4 - 5 | 861 | 66 | ||
GenBank mRNA | BC036359.1 | - | 4 - 5 | 862 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 3201 | MIR3201 | -264.5kb | Anti-sense | Chr. 22 - 48274364 - 48274415 on NCBI Build GRCh38 | |
Downstream | microRNA 4535 | MIR4535 | 235.4kb | Anti-sense | Chr. 22 - 48780295 - 48780353 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000896.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3621701 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P138628 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P138628 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 30776 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 30384 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 22585 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P138628 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3621701 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P138628 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P460740 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P138628 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 240754_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR22015007.hg.1 | ENST00000407505.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22015007.hg.1 | ENST00000467207.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22015007.hg.1 | NR_038917.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22015007.hg.1 | TC22000896.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | ENST00000625694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | ENST00000625694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | ENST00000625694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | ENST00000625694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | ENST00000625694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | ENST00000625694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | ENST00000625694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | ENST00000625694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | ENST00000626321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | ENST00000626321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | ENST00000626321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | ENST00000626321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | ENST00000626321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | ENST00000626321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | ENST00000626321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | ENST00000626321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | ENST00000630584.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | ENST00000630584.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | NR_038917.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | NR_038917.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | NR_038917.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | NR_038917.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | NR_038917.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | NR_038917.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | NR_038917.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009078.hg.1 | NR_038917.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | ENST00000625694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | ENST00000625694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | ENST00000625694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | ENST00000625694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | ENST00000625694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | ENST00000625694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | ENST00000625694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | ENST00000625694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | ENST00000626321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | ENST00000626321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | ENST00000626321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | ENST00000626321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | ENST00000626321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | ENST00000626321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | ENST00000626321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | ENST00000626321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | ENST00000630584.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | ENST00000630584.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | NR_038917.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | NR_038917.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | NR_038917.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | NR_038917.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | NR_038917.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | NR_038917.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | NR_038917.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009078.hg.1 | NR_038917.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200165616.hg.1 | TC2200009078.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200165616.hg.1 | ENST00000625694.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165616.hg.1 | ENST00000626321.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165616.hg.1 | NR_038917.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200165616.hg.1 | ENST00000625694.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165616.hg.1 | ENST00000626321.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165616.hg.1 | NR_038917.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200165614.hg.1 | TC2200009078.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200165614.hg.1 | ENST00000626321.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165614.hg.1 | ENST00000630584.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165614.hg.1 | NR_038917.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200165614.hg.1 | ENST00000626321.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165614.hg.1 | ENST00000630584.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165614.hg.1 | NR_038917.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 240754_at | - | - |
Human Genome U133 Plus 2.0 Array | 240754_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240754_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240754_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240754_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240754_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240754_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240754_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240754_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240754_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240754_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240754_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240754_at4 | - | - |
Human Genome U95 Set | 91598_PM_at | - | - |
Human Genome U95 Set | 91598_at | - | - |
Human Genome U133 Set | 240754_at | - | - |
Human Genome U133 Set | 240754_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019952 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019952 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1864749 | BC048428.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1864749 | BC048428.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1864749 | BC048428.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 7160687 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_79572 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1864749 | BC048428.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1864749 | BC048428.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1864749 | BC048428.1 | human_transcriptome |
