Assay Details
| Gene Symbol: STRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC036416.1 | - | 1 | 453 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | vitrin | VIT | -36.2kb | Anti-sense | Chr. 2 - 36696685 - 36814794 on NCBI Build GRCh38 | |
| Downstream | HEAT repeat containing 5B | HEATR5B | 128.5kb | Sense | Chr. 2 - 36980999 - 37084384 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

STRN
Genomic Map