Assay Details
Gene Symbol: LOC574538 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033859.1 | 6 - 7 | 2315 | 68 | ||
GenBank mRNA | AK096077.1 | - | 6 - 7 | 2315 | 68 | |
BC036774.1 | - | 5 - 6 | 4481 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | family with sequence similarity 138 member D | FAM138D | F379 | -102.1kb | Sense | Chr. 12 - 36661 - 38133 on NCBI Build GRCh38 |
Downstream | solute carrier family 6 member 12 | SLC6A12 |
BGT-1, BGT1, GAT2 |
46.7kb | Sense | Chr. 12 - 190077 - 214523 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001076.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3811233 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 38120 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3811233 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3811233 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P871388 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1562251_PM_a_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR12014205.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12014205.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12014205.hg.1 | TC12001076.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12014204.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12014204.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12014204.hg.1 | TC12001076.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12014208.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12014208.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12014208.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12014208.hg.1 | TC12001076.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12014206.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12014206.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12014206.hg.1 | TC12001076.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1200171273.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171273.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171273.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200171273.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171273.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171273.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200171273.hg.1 | TC1200009542.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200171274.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171274.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171274.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200171274.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171274.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171274.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200171274.hg.1 | TC1200009542.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200171278.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171278.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171278.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171278.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200171278.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171278.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171278.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171278.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200171278.hg.1 | TC1200009542.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200171272.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171272.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171272.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200171272.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171272.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171272.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200171272.hg.1 | TC1200009542.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 1562251_PM_a_at | - | - |
Human Genome U133 Plus 2.0 Array | 1562251_a_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562251_a_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562251_a_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562251_a_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562251_a_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562251_a_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562251_a_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562251_a_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562251_a_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562251_a_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562251_a_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562251_a_at7 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047172 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047172 | - | - |
