Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012837.hg.1 |
ENST00000449259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012837.hg.1 |
ENST00000449259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012837.hg.1 |
ENST00000449259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012837.hg.1 |
ENST00000449259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012837.hg.1 |
NR_036586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012837.hg.1 |
NR_036586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012837.hg.1 |
NR_036586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012837.hg.1 |
NR_036586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012837.hg.1 |
NR_036586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030621.hg.1 |
TC02001919.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030621.hg.1 |
ENST00000449259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030621.hg.1 |
NR_036586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030623.hg.1 |
TC02001919.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030623.hg.1 |
NR_036586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012837.hg.1 |
ENST00000449259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012837.hg.1 |
ENST00000449259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012837.hg.1 |
ENST00000449259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012837.hg.1 |
ENST00000449259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012837.hg.1 |
NR_036586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012837.hg.1 |
NR_036586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012837.hg.1 |
NR_036586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012837.hg.1 |
NR_036586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012837.hg.1 |
NR_036586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200193570.hg.1 |
ENST00000449259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200193570.hg.1 |
NR_036586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200193570.hg.1 |
ENST00000449259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200193570.hg.1 |
NR_036586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200193570.hg.1 |
TC0200012837.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200193573.hg.1 |
NR_036586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200193573.hg.1 |
NR_036586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200193573.hg.1 |
TC0200012837.hg.1 |
Clariom D TC |