Assay Details
Gene Symbol: CRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF335247.1 | - | 1 | 1053 | 100 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tetrapeptide repeat homeobox 1 | TPRX1 | TPRX | -16.9kb | Anti-sense | Chr. 19 - 47801243 - 47803604 on NCBI Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 1595 | LINC01595 | 41.2kb | Anti-sense | Chr. 19 - 47863229 - 47869361 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000556527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000556527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000602001.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000602001.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000602001.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000604324.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000604324.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000604324.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000604324.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000221996.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000539067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000556527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000556527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000602001.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000602001.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000602001.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000604324.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000604324.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000604324.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000604324.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | ENST00000613299.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | NM_000554.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011772.hg.1 | NM_000554.5 | Refseq |
