Assay Details
| Gene Symbol: LOC100128508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF370407.1 | - | 1 | 1251 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100130744 | LOC100130744 | -58.5kb | Anti-sense | Chr. 5 - 14712694 - 14716529 on NCBI Build GRCh38 | |
| Downstream | microRNA 4637 | MIR4637 | 48.9kb | Sense | Chr. 5 - 14825929 - 14826012 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100128508
Genomic Map