Assay Details
| Gene Symbol: KCNQ1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_130721.1 | 2 - 3 | 780 | 83 | ||
| GenBank mRNA | AJ251642.1 | - | 3 - 4 | 932 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | KCNQ1 opposite strand/antisense transcript 1 (non-protein coding) | KCNQ1OT1 |
KCNQ1-AS2, KCNQ10T1, Kncq1, KvDMR1, KvLQT1-AS, LIT1, NCRNA00012 |
-140.1kb | Sense | Chr. 11 - 2608328 - 2699998 on NCBI Build GRCh38 |
| Downstream | KCNQ1 downstream neighbor (non-protein coding) | KCNQ1DN |
BWRT, HSA404617 |
11.1kb | Anti-sense | Chr. 11 - 2870033 - 2872105 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604924 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604955 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604921 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604947 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604938 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28002076 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604944 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604943 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28002034 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28002052 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604912 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604937 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604928 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604927 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604941 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604949 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28002078 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28002044 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28002058 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28002042 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604940 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604930 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604923 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604932 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28002075 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604935 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604946 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604915 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604952 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604934 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604918 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07604925 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 151773 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 32148 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 8867 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 426662 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 67493 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 355132 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 61848 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 264979 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 167286 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 375887 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 269421 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 386083 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 215423 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 69061 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 170884 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 372889 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 395839 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3794213 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 21133 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3794213 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P02382035 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P02382035 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P02382026 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 21575 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 30368 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02382021 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02382010 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02382026 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02382016 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02382035 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19131633 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02382026 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02382035 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1562960_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009811.hg.1 | ENST00000440887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009811.hg.1 | ENST00000440887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009811.hg.1 | ENST00000440887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009811.hg.1 | ENST00000440887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009811.hg.1 | ENST00000440887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009811.hg.1 | ENST00000440887.2 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 1562960_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1562960_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562960_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562960_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562960_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562960_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562960_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562960_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562960_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562960_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562960_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562960_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562960_at9 | - | - |

KCNQ1-AS1
Genomic Map