Assay Details
| Gene Symbol: DDX50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC014263.2 | - | 1 | 651 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | storkhead box 1 | STOX1 | C10orf24 | -5.9kb | Sense | Chr. 10 - 68827537 - 68895942 on NCBI Build GRCh38 |
| Downstream | DEAD-box helicase 21 | DDX21 |
GUA, GURDB, RH-II/GU, RH-II/GuA |
53.9kb | Sense | Chr. 10 - 68956123 - 68985069 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

DDX50
Genomic Map