Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010949.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010949.hg.1 |
TC17000850.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008898.hg.1 |
ENST00000582668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008898.hg.1 |
ENST00000582668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008898.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008898.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008898.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008898.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008898.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008898.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008898.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008898.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008898.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008898.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008898.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008898.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008898.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008898.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008898.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008898.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008898.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166573.hg.1 |
ENST00000582668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166573.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166573.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166573.hg.1 |
ENST00000582668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166573.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166573.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166573.hg.1 |
TC1700008898.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700166572.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166572.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166572.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166572.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166572.hg.1 |
TC1700008898.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700166574.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166574.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166574.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166574.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166574.hg.1 |
TC1700008898.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700166566.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166566.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166566.hg.1 |
TC1700008898.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700008898.hg.1 |
ENST00000582668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008898.hg.1 |
ENST00000582668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008898.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008898.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008898.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008898.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008898.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008898.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008898.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008898.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008898.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008898.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008898.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008898.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008898.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008898.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008898.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008898.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008898.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166567.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166567.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166567.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166567.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166567.hg.1 |
TC1700008898.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700166568.hg.1 |
ENST00000582668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166568.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166568.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166568.hg.1 |
ENST00000582668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166568.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166568.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166568.hg.1 |
TC1700008898.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700166569.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166569.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166569.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166569.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166569.hg.1 |
TC1700008898.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700166571.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166571.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166571.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166571.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166571.hg.1 |
TC1700008898.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700166570.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166570.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166570.hg.1 |
ENST00000585075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166570.hg.1 |
NR_036520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166570.hg.1 |
TC1700008898.hg.1 |
Clariom D TC |