Assay Details
| Gene Symbol: LUC7L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC041001.2 | - | 1 | 3149 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | C7orf55-LUC7L2 readthrough | C7orf55-LUC7L2 | Sense | Chr. 7 - 139341132 - 139423457 on NCBI Build GRCh38 | ||
| Downstream | killer cell lectin like receptor G2 | KLRG2 | CLEC15B | 3.5kb | Anti-sense | Chr. 7 - 139426931 - 139483716 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LUC7L2
Genomic Map