| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007195.hg.1 |
ENST00000377680.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007195.hg.1 |
ENST00000377680.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007195.hg.1 |
ENST00000377680.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007195.hg.1 |
ENST00000377680.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007195.hg.1 |
ENST00000471864.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007195.hg.1 |
ENST00000471864.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007195.hg.1 |
ENST00000484285.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007195.hg.1 |
ENST00000484285.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007195.hg.1 |
ENST00000484285.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007195.hg.1 |
NR_027294.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007195.hg.1 |
NR_027294.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007195.hg.1 |
NR_027294.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007195.hg.1 |
ENST00000377680.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007195.hg.1 |
ENST00000377680.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007195.hg.1 |
ENST00000377680.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007195.hg.1 |
ENST00000377680.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007195.hg.1 |
ENST00000471864.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007195.hg.1 |
ENST00000471864.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007195.hg.1 |
ENST00000484285.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007195.hg.1 |
ENST00000484285.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007195.hg.1 |
ENST00000484285.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007195.hg.1 |
NR_027294.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007195.hg.1 |
NR_027294.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007195.hg.1 |
NR_027294.1 |
Refseq |