Assay Details
| Gene Symbol: LOC146513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038438.1 | 1 - 2 | 112 | 77 | ||
| GenBank mRNA | BC043214.2 | - | 1 - 2 | 116 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1081 | LINC01081 |
TCONS00024764, TCONS_00024764 |
-180.0 | Anti-sense | Chr. 16 - 86225580 - 86286247 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 917 | LINC00917 | 41.1kb | Anti-sense | Chr. 16 - 86331850 - 86345679 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000667.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711896 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711766 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169169 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169106 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169179 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711889 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169137 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169158 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169079 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169077 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711891 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169088 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711832 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169192 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16922739 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169184 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169092 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169072 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169121 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169083 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169132 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711906 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169164 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169128 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169085 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169173 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711848 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169180 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169175 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169134 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711909 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169189 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711754 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169080 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711853 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169107 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169111 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169196 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169172 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169167 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711800 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16922775 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711820 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169135 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711844 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169176 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169171 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169129 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711838 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711894 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711819 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169067 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711789 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711858 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169160 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169102 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169099 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169197 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711856 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711753 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711816 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169181 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711845 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711847 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169078 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169190 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169153 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169097 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169163 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169108 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711791 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711846 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169133 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16922751 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169094 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16922774 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711836 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169202 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169074 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169142 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711831 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711890 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169125 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711774 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711758 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169165 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711837 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169170 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169161 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711805 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169124 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711756 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169062 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169156 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711812 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711839 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169112 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711778 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711911 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169093 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711773 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711777 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16922757 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711804 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169118 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169076 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711822 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169162 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711779 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169178 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169199 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711787 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169122 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169120 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169082 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169174 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169095 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169193 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711874 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169194 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16922772 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711807 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16922780 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169113 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16922777 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169168 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10169136 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711794 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30711815 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 8346 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 647699 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 637937 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 420225 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 17742 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3816280 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 14412 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3816280 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3816280 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P20550397 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 160054 | BC032422.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 160054 | BC050730.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 160054 | L76662.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 160054 | U50546.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 160054 | U73395.1 | genbank_mrna_human |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P12545853 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03194263 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20550397 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P20550397 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1562623_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009445.hg.1 | ENST00000511252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009445.hg.1 | NR_038438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009445.hg.1 | TC16000667.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009446.hg.1 | ENST00000511252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009446.hg.1 | NR_038438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009446.hg.1 | TC16000667.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009447.hg.1 | ENST00000511252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009447.hg.1 | NR_038438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009447.hg.1 | TC16000667.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008726.hg.1 | ENST00000599486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008726.hg.1 | ENST00000599486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008726.hg.1 | ENST00000599486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008726.hg.1 | NR_038438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008726.hg.1 | NR_038438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008726.hg.1 | NR_038438.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600165192.hg.1 | ENST00000599486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600165192.hg.1 | NR_038438.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600165192.hg.1 | ENST00000599486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600165192.hg.1 | NR_038438.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600165192.hg.1 | TC1600008726.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600165190.hg.1 | ENST00000599486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600165190.hg.1 | NR_038438.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600165190.hg.1 | ENST00000599486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600165190.hg.1 | NR_038438.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600165190.hg.1 | TC1600008726.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600165191.hg.1 | ENST00000599486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600165191.hg.1 | NR_038438.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600165191.hg.1 | ENST00000599486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600165191.hg.1 | NR_038438.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600165191.hg.1 | TC1600008726.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600008726.hg.1 | ENST00000599486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008726.hg.1 | ENST00000599486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008726.hg.1 | ENST00000599486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008726.hg.1 | NR_038438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008726.hg.1 | NR_038438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008726.hg.1 | NR_038438.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 1562623_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1562623_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562623_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562623_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562623_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562623_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562623_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562623_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562623_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562623_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562623_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562623_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562623_at9 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054873 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054873 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina human-6 v2.0 expression beadchip | ILMN_1843203 | - | - |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 4060678 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_91184 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1843203 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1843203 | - | - |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1843203 | - | - |

LOC146513
Genomic Map