Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009096.hg.1 |
ENST00000412672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009096.hg.1 |
ENST00000412672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009096.hg.1 |
ENST00000412672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009096.hg.1 |
ENST00000412672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009096.hg.1 |
NR_027124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009096.hg.1 |
NR_027124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009096.hg.1 |
NR_027124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009096.hg.1 |
NR_027124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011563.hg.1 |
ENST00000412672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011563.hg.1 |
NR_027124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011563.hg.1 |
TC20000843.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011564.hg.1 |
ENST00000412672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011564.hg.1 |
NR_027124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011564.hg.1 |
TC20000843.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000163701.hg.1 |
ENST00000412672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000163701.hg.1 |
NR_027124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000163701.hg.1 |
TC2000009096.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000163701.hg.1 |
ENST00000412672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000163701.hg.1 |
NR_027124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009096.hg.1 |
ENST00000412672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009096.hg.1 |
ENST00000412672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009096.hg.1 |
ENST00000412672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009096.hg.1 |
ENST00000412672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009096.hg.1 |
NR_027124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009096.hg.1 |
NR_027124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009096.hg.1 |
NR_027124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009096.hg.1 |
NR_027124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000163702.hg.1 |
ENST00000412672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000163702.hg.1 |
NR_027124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000163702.hg.1 |
TC2000009096.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000163702.hg.1 |
ENST00000412672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000163702.hg.1 |
NR_027124.1 |
Refseq |