Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001470.hg.1 |
TC05000145.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001470.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001470.hg.1 |
ENST00000507693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001470.hg.1 |
NR_033832.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000507693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000507693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000507693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000507693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
ENST00000507693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
NR_033832.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
NR_033832.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
NR_033832.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
NR_033832.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
NR_033832.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
NR_033832.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
NR_033832.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
NR_033832.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007081.hg.1 |
NR_033832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000507693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000507693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000507693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000507693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
ENST00000507693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
NR_033832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
NR_033832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
NR_033832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
NR_033832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
NR_033832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
NR_033832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
NR_033832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
NR_033832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007081.hg.1 |
NR_033832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500147801.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500147801.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500147801.hg.1 |
ENST00000507693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500147801.hg.1 |
NR_033832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500147801.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500147801.hg.1 |
ENST00000505518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500147801.hg.1 |
ENST00000507693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500147801.hg.1 |
NR_033832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500147801.hg.1 |
TC0500007081.hg.1 |
Clariom D TC |