Assay Details
Gene Symbol: LOC100126784 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_015384.2 | 1 | 3619 | 80 | ||
GenBank mRNA | BC047636.1 | - | 1 | 1449 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 4486 | MIR4486 | -135.6kb | Anti-sense | Chr. 11 - 19575310 - 19575372 on NCBI Build GRCh38 | |
Downstream | microRNA 4694 | MIR4694 | 47.5kb | Sense | Chr. 11 - 19760004 - 19760083 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3262083 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P187651 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 1321 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 11209 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3262083 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3262083 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677372 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677416 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077727 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077704 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077721 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077724 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077731 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677388 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677365 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677399 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677385 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677402 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677409 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677382 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677375 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077697 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077720 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077705 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077702 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677370 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077688 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077729 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677378 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077715 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677422 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077703 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077710 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677397 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677420 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077717 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077701 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677362 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077686 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077714 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677368 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077722 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677380 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677360 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677361 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677403 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677358 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077722 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077717 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077724 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077703 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077697 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077721 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677375 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077714 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077702 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077705 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677397 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677361 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677388 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077686 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677358 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677380 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677399 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077710 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677403 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677378 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077727 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677409 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077701 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677382 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077729 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677372 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077704 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677360 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077731 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077688 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677365 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677362 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677385 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677402 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677420 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677422 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677368 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677416 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07677370 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077715 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28077720 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P187651 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P187651 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P187654 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P518251 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677362 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677403 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677342 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677350 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677361 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677387 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677372 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677382 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677341 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677398 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677416 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677380 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677421 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677370 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677378 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677368 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677366 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677409 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677405 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677426 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677396 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677388 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677345 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677400 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677419 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16426123 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677364 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677375 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677424 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677406 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677392 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677411 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677379 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677358 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677414 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677351 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677428 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677353 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677347 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07677385 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 240407_PM_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 236102_PM_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 239141_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR11018745.hg.1 | NR_015384.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11018745.hg.1 | TC11001483.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC11001483.hg.1 | NR_015384.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178359.hg.1 | NR_015384.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178359.hg.1 | NR_015384.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178359.hg.1 | TC1100010301.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100010301.hg.1 | ENST00000603468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010301.hg.1 | NR_015384.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010301.hg.1 | NR_015384.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010301.hg.1 | NR_015384.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010301.hg.1 | NR_015384.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010301.hg.1 | NR_015384.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010301.hg.1 | NR_015384.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178362.hg.1 | NR_015384.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178362.hg.1 | NR_015384.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178362.hg.1 | TC1100010301.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100178363.hg.1 | NR_015384.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178363.hg.1 | NR_015384.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178363.hg.1 | TC1100010301.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100178358.hg.1 | ENST00000603468.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178358.hg.1 | NR_015384.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178358.hg.1 | ENST00000603468.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178358.hg.1 | NR_015384.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178358.hg.1 | TC1100010301.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100178360.hg.1 | NR_015384.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178360.hg.1 | NR_015384.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178360.hg.1 | TC1100010301.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100178361.hg.1 | NR_015384.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178361.hg.1 | NR_015384.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178361.hg.1 | TC1100010301.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 240407_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 239141_at | - | - |
Human Genome U133 Plus 2.0 Array | 236102_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 239141_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 240407_at | - | - |
Human Genome U133 Plus 2.0 Array | 236102_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239141_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 236102_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240407_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240407_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 236102_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239141_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239141_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239141_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239141_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239141_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240407_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 236102_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 236102_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240407_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240407_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239141_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240407_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240407_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 236102_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239141_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 236102_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 236102_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239141_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240407_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239141_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 236102_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 236102_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239141_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240407_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 236102_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240407_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 236102_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240407_at5 | - | - |
Human Genome U95 Set | 50991_at | - | - |
Human Genome U95 Set | 50991_PM_at | - | - |
Human Genome U95 Set | 64731_r_at | - | - |
Human Genome U95 Set | 64731_PM_r_at | - | - |
Human Genome U95 Set | 87208_at | - | - |
Human Genome U95 Set | 87208_PM_at | - | - |
Human Genome U95 Set | 87207_at | - | - |
Human Genome U95 Set | 87207_PM_at | - | - |
Human Genome U133 Set | 239141_at | - | - |
Human Genome U133 Set | 239141_PM_at | - | - |
Human Genome U133 Set | 236102_at | - | - |
Human Genome U133 Set | 240407_PM_at | - | - |
Human Genome U133 Set | 236102_PM_at | - | - |
Human Genome U133 Set | 240407_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A045010 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A045010 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 229493 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3268883 | BC035903.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3268883 | NR_015384.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3268883 | BC029949.1 | human_transcriptome |
