Assay Details
Gene Symbol: HTR5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024012.3 | NP_076917.1 | 2 | 4341 | 73 | |
GenBank mRNA | BX538209.1 | - | 1 | 1655 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | HTR5A antisense RNA 1 | HTR5A-AS1 | HTR5AOS | -14.0kb | Anti-sense | Chr. 7 - 155067067 - 155071557 on NCBI Build GRCh38 |
Downstream | insulin induced gene 1 | INSIG1 |
CL-6, CL6 |
210.4kb | Sense | Chr. 7 - 155297776 - 155310235 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
