Assay Details
| Gene Symbol: LOC284454 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036515.1 | 1 | 446 | 67 | ||
| GenBank mRNA | AL832183.1 | - | 1 | 445 | 67 | |
| BX640708.1 | - | 1 | 446 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 24-2 | MIR24-2 |
MIRN24-2, miR-24-2, miRNA24-2 |
Sense | Chr. 19 - 13836287 - 13836359 on NCBI Build GRCh38 | |
| Downstream | microRNA 27a | MIR27A |
MIR27, MIRN27A, mir-27a |
151.0 | Sense | Chr. 19 - 13836440 - 13836517 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001224.hg.1
Set Membership: |
|

LOC284454
Genomic Map