Assay Details
| Gene Symbol: TNC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX641111.1 | - | 1 | 4554 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tumor necrosis factor superfamily member 8 | TNFSF8 |
CD153, CD30L, CD30LG, TNLG3A |
-116.6kb | Sense | Chr. 9 - 114893343 - 114930595 on NCBI Build GRCh38 |
| Downstream | deleted in esophageal cancer 1 | DEC1 | CTS9 | 59.8kb | Anti-sense | Chr. 9 - 115141818 - 115402644 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TNC
Genomic Map