Assay Details
Gene Symbol: GATA6-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_102763.1 | 1 - 2 | 301 | 87 | ||
GenBank mRNA | AK123079.1 | - | 1 - 2 | 301 | 87 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | mindbomb E3 ubiquitin protein ligase 1 | MIB1 |
DIP-1, DIP1, LVNC7, MIB, ZZANK2, ZZZ6 |
-295.9kb | Anti-sense | Chr. 18 - 21740793 - 21870957 on NCBI Build GRCh38 |
Downstream | GATA binding protein 6 | GATA6 | 469.0 | Anti-sense | Chr. 18 - 22169437 - 22202528 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31164158 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31164152 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31164148 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10565386 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10565379 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10565384 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 808471 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P125832 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 7984 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 29027 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P125832 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31164152 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31164156 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31164159 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10565386 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10565380 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10565384 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10565381 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31164148 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10565388 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31164147 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10565385 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10565379 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31164158 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10565385 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31164158 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31164156 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31164152 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10565384 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31164148 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10565381 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31164147 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10565388 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10565379 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10565380 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31164159 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10565386 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P125832 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P20788507 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20788507 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P125832 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P20788507 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 236118_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000579431.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000579431.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000579431.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000579431.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000579431.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000579431.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000584201.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000584201.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000584373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000584373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000584373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000584373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | ENST00000584373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156306.hg.1 | ENST00000579431.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156306.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156306.hg.1 | ENST00000584373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156306.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156306.hg.1 | TC1800008249.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800156306.hg.1 | ENST00000579431.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156306.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156306.hg.1 | ENST00000584373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156306.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156308.hg.1 | ENST00000579431.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156308.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156308.hg.1 | ENST00000584373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156308.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156308.hg.1 | TC1800008249.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800156308.hg.1 | ENST00000579431.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156308.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156308.hg.1 | ENST00000584373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156308.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156304.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156304.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156304.hg.1 | TC1800008249.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800156304.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156304.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156311.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156311.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156311.hg.1 | TC1800008249.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800156311.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156311.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156317.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156317.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156317.hg.1 | TC1800008249.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800156317.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156317.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156305.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156305.hg.1 | ENST00000584373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156305.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156305.hg.1 | TC1800008249.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800156305.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156305.hg.1 | ENST00000584373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156305.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156307.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156307.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156307.hg.1 | TC1800008249.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800156307.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156307.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156309.hg.1 | ENST00000579431.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156309.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156309.hg.1 | ENST00000584201.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156309.hg.1 | ENST00000584373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156309.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156309.hg.1 | TC1800008249.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800156309.hg.1 | ENST00000579431.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156309.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156309.hg.1 | ENST00000584201.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156309.hg.1 | ENST00000584373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156309.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156314.hg.1 | ENST00000579431.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156314.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156314.hg.1 | ENST00000584201.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156314.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156314.hg.1 | TC1800008249.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800156314.hg.1 | ENST00000579431.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156314.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156314.hg.1 | ENST00000584201.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156314.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000579431.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000579431.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000579431.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000579431.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000579431.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000579431.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000584201.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000584201.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000584373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000584373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000584373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000584373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | ENST00000584373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008249.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156313.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156313.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156313.hg.1 | TC1800008249.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800156313.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156313.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156310.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156310.hg.1 | ENST00000584373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156310.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156310.hg.1 | TC1800008249.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800156310.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156310.hg.1 | ENST00000584373.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156310.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156312.hg.1 | ENST00000579431.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156312.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156312.hg.1 | NR_102763.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800156312.hg.1 | TC1800008249.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800156312.hg.1 | ENST00000579431.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156312.hg.1 | ENST00000583490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800156312.hg.1 | NR_102763.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 236118_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 236118_at | - | - |
Human Genome U95 Set | 56806_at | - | - |
Human Genome U95 Set | 56806_PM_at | - | - |
Human Genome U95 Set | 89731_at | - | - |
Human Genome U95 Set | 89731_PM_at | - | - |
Human Genome U133 Set | 236118_at | - | - |
Human Genome U133 Set | 236118_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 248820 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3178013 | AK123079.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3254481 | AK123079.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1816925 | AK123079.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 3460739 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_98728 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1816925 | AK123079.1 | human_transcriptome |
