Assay Details
Gene Symbol: FLJ41130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK123125.1 | - | 1 - 2 | 1153 | 93 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | phosphatidylinositol glycan anchor biosynthesis class G | PIGG |
GPI7, LAS21, MRT53, PRO4405, RLGS1930 |
-37.2kb | Sense | Chr. 4 - 499200 - 539921 on NCBI Build GRCh38 |
Downstream | phosphodiesterase 6B | PDE6B |
CSNB3, CSNBAD2, PDEB, RP40, rd1 |
8.5kb | Sense | Chr. 4 - 587593 - 670892 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886340 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030924 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886357 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886385 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15530829 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030938 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886343 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886369 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030905 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886309 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886384 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886312 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886315 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886305 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030944 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886374 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886338 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030907 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886348 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886336 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886316 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886293 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030893 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030917 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030920 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886356 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23031014 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23031009 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886363 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886360 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030909 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030940 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886368 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15530830 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886371 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030923 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030976 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886319 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886361 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886300 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886294 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886286 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030949 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030911 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15530837 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886383 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030931 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886314 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030992 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030945 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886382 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15530840 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886295 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030947 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886380 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886290 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886289 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886298 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886332 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886330 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15530832 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030946 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030969 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886379 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886292 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886367 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030932 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886337 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886375 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030913 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886344 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886299 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886329 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23030927 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886372 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02886364 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 159243 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 896144 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 111403 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3525377 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 15329 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P36614329 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3525377 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P36614329 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 4898 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P14578709 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00906283 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P36614329 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P36614329 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1912611 | AK123125.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1912611 | AK123125.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1912611 | AK123125.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_99435 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_99435 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1912611 | AK123125.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1912611 | AK123125.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1912611 | AK123125.1 | human_transcriptome |
