Assay Details
Gene Symbol: JAKMIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK123426.1 | - | 1 | 1250 | 90 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | BCL2 interacting protein 3 | BNIP3 | NIP3 | -160.2kb | Anti-sense | Chr. 10 - 131967683 - 131982013 on NCBI Build GRCh38 |
Downstream | dihydropyrimidinase like 4 | DPYSL4 |
CRMP3, DRP-4, ULIP4 |
40.2kb | Sense | Chr. 10 - 132184983 - 132205776 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
