Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012467.hg.1 |
ENST00000581549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012467.hg.1 |
ENST00000581549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012467.hg.1 |
ENST00000581549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012467.hg.1 |
ENST00000581549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012467.hg.1 |
ENST00000581549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012467.hg.1 |
ENST00000581549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012467.hg.1 |
ENST00000581549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012467.hg.1 |
NR_033876.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012467.hg.1 |
NR_033876.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012467.hg.1 |
NR_033876.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012467.hg.1 |
NR_033876.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012467.hg.1 |
NR_033876.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012467.hg.1 |
NR_033876.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012467.hg.1 |
NR_033876.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024740.hg.1 |
ENST00000499674.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024740.hg.1 |
NR_033876.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024740.hg.1 |
TC17001841.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700207819.hg.1 |
ENST00000581549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700207819.hg.1 |
NR_033876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700207819.hg.1 |
ENST00000581549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700207819.hg.1 |
NR_033876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700207819.hg.1 |
TC1700012467.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700012467.hg.1 |
ENST00000581549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012467.hg.1 |
ENST00000581549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012467.hg.1 |
ENST00000581549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012467.hg.1 |
ENST00000581549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012467.hg.1 |
ENST00000581549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012467.hg.1 |
ENST00000581549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012467.hg.1 |
ENST00000581549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012467.hg.1 |
NR_033876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012467.hg.1 |
NR_033876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012467.hg.1 |
NR_033876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012467.hg.1 |
NR_033876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012467.hg.1 |
NR_033876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012467.hg.1 |
NR_033876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012467.hg.1 |
NR_033876.1 |
Refseq |